missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-oxo-pentanoate, 99+%, Thermo Scientific Chemicals

Catalog Number p-166748
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
30414-53-0
C6H10O3
130.14
MFCD00011705
XJMIXEAZMCTAGH-UHFFFAOYSA-N
methyl 3-oxovalerate, methyl propionylacetate, methyl 3-oxo-pentanoate, pentanoic acid, 3-oxo-, methyl ester, methyl-3-oxo pentanoate, methyl 3-oxo-n-valerate, valeric acid, 3-oxo-, methyl ester, 3-oxovaleric acid methyl ester, 3-oxo-pentanoic acid methyl ester, 3-keto-n-valeric acid methyl ester
121699
methyl 3-oxopentanoate
CCC(=O)CC(=O)OC
This item is not returnable. View return policy
30414-53-0
C6H10O3
130.14
MFCD00011705
XJMIXEAZMCTAGH-UHFFFAOYSA-N
methyl 3-oxovalerate, methyl propionylacetate, methyl 3-oxo-pentanoate, pentanoic acid, 3-oxo-, methyl ester, methyl-3-oxo pentanoate, methyl 3-oxo-n-valerate, valeric acid, 3-oxo-, methyl ester, 3-oxovaleric acid methyl ester, 3-oxo-pentanoic acid methyl ester, 3-keto-n-valeric acid methyl ester
121699
methyl 3-oxopentanoate
CCC(=O)CC(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -35°C
Color Colorless to Yellow
Density 1.0300g/mL
Boiling Point 95°C to 97°C (48.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3CH2COCH2COOCH3
Refractive Index 1.4220 to 1.424
Quantity 25 mL
Fieser 10,415; 01,1022; 04,437; 05,589; 07,324; 11,472; 12,126; 16,303
Specific Gravity 1.03
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with most common organic solvents
Formula Weight 130.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Methyl 3-oxo-pentanoate, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'