missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3-hydroxy-4-iodobenzoate, 97%

Brand:  Alfa Aesar™ H64354.06

Code : NEW

Additional Details : CAS Number : 157942-12-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15441519

  • £265.80 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Description

Description

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Specifications

Chemical Identifiers

Methyl 3-hydroxy-4-iodobenzoate
278.05
7289690
157942-12-6
MFCD06203960
3-Hydroxy-4-iodobenzoic acid methyl ester

Specifications

Methyl 3-hydroxy-4-iodobenzoate
5g
Solid
278.05
7289690
Light Sensitive
157942-12-6
97%
MFCD06203960
3-Hydroxy-4-iodobenzoic acid methyl ester
Videos
SDS