missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-fluorophenylacetate, 98%, Thermo Scientific Chemicals

Catalog Number p-163287
18.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
64123-77-9
C9H9FO2
168.17
MFCD03093288
OXYLCGRXCQARQV-UHFFFAOYSA-N
methyl 2-3-fluorophenyl acetate, methyl 3-fluorophenylacetate, methyl3-fluorophenylacetate, methyl 3-fluorophenyl acetate, 3-fluorobenzeneacetic acid methyl ester, benzeneacetic acid, 3-fluoro-, methyl ester, 3-fluorophenylacetic acid methyl ester, 3-fluorophenyl acetic acid methyl ester, 3-fluoro-phenyl-acetic acid methyl ester, 2-3-fluorophenyl acetic acid methyl ester
2734108
methyl 2-(3-fluorophenyl)acetate
COC(=O)CC1=CC=CC(F)=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
64123-77-9
C9H9FO2
168.17
MFCD03093288
OXYLCGRXCQARQV-UHFFFAOYSA-N
methyl 2-3-fluorophenyl acetate, methyl 3-fluorophenylacetate, methyl3-fluorophenylacetate, methyl 3-fluorophenyl acetate, 3-fluorobenzeneacetic acid methyl ester, benzeneacetic acid, 3-fluoro-, methyl ester, 3-fluorophenylacetic acid methyl ester, 3-fluorophenyl acetic acid methyl ester, 3-fluoro-phenyl-acetic acid methyl ester, 2-3-fluorophenyl acetic acid methyl ester
2734108
methyl 2-(3-fluorophenyl)acetate
COC(=O)CC1=CC=CC(F)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH2CO2H
Refractive Index 1.4870 to 1.489
Quantity 5 g
Formula Weight 168.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 3-fluorophenylacetate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'