missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3-fluoro-4-nitrobenzoate, 97%

Brand:  Alfa Aesar™ H64688.22

Code : NEW

Additional Details : CAS Number : 185629-31-6 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15411999

  • £355.00 / 100g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 3-fluoro-4-nitrobenzoate
199.14
10657755
185629-31-6
MFCD08444027
3-Fluoro-4-nitrobenzoic acid methyl ester

Specifications

Methyl 3-fluoro-4-nitrobenzoate
185629-31-6
97%
MFCD08444027
3-Fluoro-4-nitrobenzoic acid methyl ester
100g
Solid
199.14
10657755
Videos
SDS