missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3-chloropyrazine-2-carboxylate, ≥97%

Brand:  Alfa Aesar™ H64508.03

Code : NEW

Additional Details : CAS Number : 27825-21-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15491749

  • £160.06 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 3-chloropyrazine-2-carboxylate
172.57
127506
27825-21-4
MFCD03093326
3-Chloropyrazine-2-carboxylic acid methyl ester

Specifications

Methyl 3-chloropyrazine-2-carboxylate
1g
Solid
172.57
127506
34° to 35°C
27825-21-4
≥97%
MFCD03093326
3-Chloropyrazine-2-carboxylic acid methyl ester
Videos
SDS