missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3-chloro-4-hydroxybenzoate, 98%

Brand:  Alfa Aesar™ A12389.14

Code : NEW

Additional Details : CAS Number : 3964-57-6 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11409553

  • £95.30 / 25g


Estimated Shipment date
from Supplier 25-04-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 3-chloro-4-hydroxybenzoate
C8H7ClO3
MFCD00016418
3-Chloro-4-hydroxybenzoic acid methyl ester
3964-57-6
186.6
2641291

Specifications

Methyl 3-chloro-4-hydroxybenzoate
98%
186.6
2641291
104° to 109°C
3964-57-6
C8H7ClO3
MFCD00016418
3-Chloro-4-hydroxybenzoic acid methyl ester
25g
Videos
SDS