missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3-bromopyrazine-2-carboxylate, 97%

Brand:  Alfa Aesar™ H63515.06

Code : NEW

Additional Details : CAS Number : 51171-02-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15410259

  • £717.62 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 3-bromopyrazine-2-carboxylate
217.02
3-Bromopyrazine-2-carboxylic acid methyl ester
51171-02-9
MFCD08753935

Specifications

Methyl 3-bromopyrazine-2-carboxylate
Solid
217.02
3-Bromopyrazine-2-carboxylic acid methyl ester
51171-02-9
97%
MFCD08753935
5g
Videos
SDS