missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-bromo-4-methylbenzoate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It is a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a pharmaceutical intermediate.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store at room temperature. Incompatible with oxidizing agents.
It is a pharmaceutical intermediate.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Quantity | 25 g |
Solubility Information | Soluble in dimethyl sulfoxide. |
Formula Weight | 229.07 |
Percent Purity | 99% |
Chemical Name or Material | Methyl 3-bromo-4-methylbenzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'