missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3-amino-4-methoxybenzoate, 97%

Brand:  Alfa Aesar™ A12052.14

Code : NEW

Additional Details : CAS Number : 24812-90-6 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11449932

  • £132.20 / 25g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 3-amino-4-methoxybenzoate
C9H11NO3
MFCD00017196
3-Amino-4-methoxybenzoic acid methyl ester
24812-90-6
181.19
2968002

Specifications

Methyl 3-amino-4-methoxybenzoate
97%
181.19
2968002
84° to 86°C
24812-90-6
C9H11NO3
MFCD00017196
3-Amino-4-methoxybenzoic acid methyl ester
25g
Videos
SDS