missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3,6-dibromopyrazine-2-carboxylate, 97%

Brand:  Alfa Aesar™ H63319.03

Code : NEW

Additional Details : CAS Number : 13301-04-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459958

  • £71.76 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 3,6-dibromopyrazine-2-carboxylate
295.92
3,6-Dibromopyrazine-2-carboxylic acid methyl ester
13301-04-7
MFCD11976863

Specifications

Methyl 3,6-dibromopyrazine-2-carboxylate
1g
Solid
295.92
3,6-Dibromopyrazine-2-carboxylic acid methyl ester
Off-white to brown
13301-04-7
97%
MFCD11976863
Videos
SDS