missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3,5-dichlorobenzoate, Tech., Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Assay Percent Range | 90% |
Packaging | Amber glass bottle |
Quantity | 10 g |
Formula Weight | 205.04 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | Methyl 3, 5-dichlorobenzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'