missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3-(4-chlorosulfonyl)phenylpropionate, 97%

Brand:  Alfa Aesar™ L17609.03

Code : NEW

Additional Details : CAS Number : 374537-95-8 Weight : 0.00100kg Transport : UN number : 3261 Chem class : 8 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 11338556

  • £20.99 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 3-(4-chlorosulfonyl)phenylpropionate
C10H11ClO4S
MFCD02093942
374537-95-8
262.71
UN3261

Specifications

Methyl 3-(4-chlorosulfonyl)phenylpropionate
97%
262.71
UN3261
1g
374537-95-8
C10H11ClO4S
MFCD02093942
64° to 65°C
Videos
SDS