missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-(2-morpholin-4-ylethoxy)benzoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 265.31 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 3-(2-morpholin-4-ylethoxy)benzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'