missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 2-(trifluoromethyl)isonicotinate, 95%

Brand:  Alfa Aesar™ H63273.03

Code : NEW

Additional Details : CAS Number : 588702-68-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15479888

  • £208.82 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 2-(trifluoromethyl)isonicotinate
205.13
2-(Trifluoromethyl)isonicotinic acid methyl ester; Methyl 2-(trifluoromethyl)pyridine-4-carboxylate
588702-68-5
MFCD14698093

Specifications

Methyl 2-(trifluoromethyl)isonicotinate
1g
Liquid
205.13
2-(Trifluoromethyl)isonicotinic acid methyl ester; Methyl 2-(trifluoromethyl)pyridine-4-carboxylate
588702-68-5
Colorless
95%
MFCD14698093
Videos
SDS