missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-oxocyclohexanecarboxylate, 90%, Thermo Scientific Chemicals

Catalog Number p-3523491
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
41302-34-5
C8H12O3
156.2
MFCD00077699
JEENWEAPRWGXSG-UHFFFAOYSA-N
methyl 2-oxocyclohexanecarboxylate, 2-methoxycarbonylcyclohexanone, 2-carbomethoxycyclohexanone, cyclohexanecarboxylic acid, 2-oxo-, methyl ester, methyl cyclohexanone-2-carboxylate, 2-oxo-cyclohexanecarboxylic acid methyl ester, 2-hydroxy methyl cyclohexanone, 2-oxocyclohexanecarboxylic acid, methyl ester, pubchem20380, acmc-1ao3u
170489
methyl 2-oxocyclohexane-1-carboxylate
COC(=O)C1CCCCC1=O
This item is not returnable. View return policy
41302-34-5
C8H12O3
156.2
MFCD00077699
JEENWEAPRWGXSG-UHFFFAOYSA-N
methyl 2-oxocyclohexanecarboxylate, 2-methoxycarbonylcyclohexanone, 2-carbomethoxycyclohexanone, cyclohexanecarboxylic acid, 2-oxo-, methyl ester, methyl cyclohexanone-2-carboxylate, 2-oxo-cyclohexanecarboxylic acid methyl ester, 2-hydroxy methyl cyclohexanone, 2-oxocyclohexanecarboxylic acid, methyl ester, pubchem20380, acmc-1ao3u
170489
methyl 2-oxocyclohexane-1-carboxylate
COC(=O)C1CCCCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1090g/mL
Boiling Point 50°C (0.2 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. may occur as tautomers (GC)
Packaging Glass bottle
Refractive Index 1.4830 to 1.486
Quantity 10 mL
Beilstein 10, IV, 2606
Specific Gravity 1.109
Formula Weight 156.2
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Methyl 2-oxocyclohexanecarboxylate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'