missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(methylthio)butyrate, 98%, Thermo Scientific™

Catalog Number p-7026345
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
51534-66-8
C6H12O2S
148.22
MFCD00013393
PBYSEUNXLXTEAN-UHFFFAOYSA-N
560572
methyl 2-methylsulfanylbutanoate
CCC(C(=O)OC)SC
This item is not returnable. View return policy
51534-66-8
C6H12O2S
148.22
MFCD00013393
PBYSEUNXLXTEAN-UHFFFAOYSA-N
560572
methyl 2-methylsulfanylbutanoate
CCC(C(=O)OC)SC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.028
Boiling Point 103°C to 104°C (35 mmHg)
Flash Point 64°C (147°F)
Refractive Index 1.461
Quantity 5 g
Beilstein 1925141
Synonym methyl 2-methylthio butyrate, methyl 2-methylsulfanyl butanoate, butanoic acid,2-methylthio-, methyl ester, methyl 2-methylthio-butyrate, methyl alpha-methylsulfanyl butyrate, acmc-20akj9, 2-methylthio-butanoic acid methyl ester, butanoic acid, 2-methylthio-, methyl ester, +/--butanoic acid, 2-methylthio-, methyl ester, butanoic acid, 2-methylthio-, methyl ester, +/-
Formula Weight 148.23
Percent Purity 98%
Chemical Name or Material Methyl 2-(methylthio)butyrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'