missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methylnicotinate, 97%, Thermo Scientific Chemicals

Catalog Number 13581630
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
65719-09-7
C8H9NO2
151.16
KLHWBYHFWALOIJ-UHFFFAOYSA-N
7015327
methyl 2-methylpyridine-3-carboxylate
CC1=C(C=CC=N1)C(=O)OC
This item is not returnable. View return policy
65719-09-7
C8H9NO2
151.16
KLHWBYHFWALOIJ-UHFFFAOYSA-N
7015327
methyl 2-methylpyridine-3-carboxylate
CC1=C(C=CC=N1)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 102.0°C to 104.0°C (10.0 mmHg)
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Synonym methyl 2-methylnicotinate, 2-methylnicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2-methyl-, methyl ester, methyl2-methylnicotinate, pubchem2218, acmc-1belg, methyi 2-methyinicotinate, methyl 2-methyinicotinate, methyl-2-methylnicotinate, ksc493g0t
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material Methyl 2-methylnicotinate
compliance-icons
Product Identifier
  • Methyl 2-methylnicotinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'