missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hydroxynicotinate, 97%, Thermo Scientific Chemicals

Catalog Number 15406147
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
10128-91-3
C7H7NO3
153.137
MFCD00661282
SILBTMNCGYLTOK-UHFFFAOYSA-N
methyl 2-hydroxynicotinate, methyl 2-oxo-1,2-dihydro-3-pyridinecarboxylate, methyl 2-oxo-1,2-dihydropyridine-3-carboxylate, methyl 2-hydroxypyridine-3-carboxylate, 2-hydroxy-nicotinic acid methyl ester, 2-hydroxynicotinic acid methyl ester, 2-hydroxy-3-pyridinecarboxylic acid methyl ester, 3-pyridinecarboxylic acid, 2-hydroxy-, methyl ester, methyl 2-hydroxy-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 1,2-dihydro-2-oxo-, methyl ester
579028
methyl 2-oxo-1H-pyridine-3-carboxylate
COC(=O)C1=CC=CNC1=O
This item is not returnable. View return policy
10128-91-3
C7H7NO3
153.137
MFCD00661282
SILBTMNCGYLTOK-UHFFFAOYSA-N
methyl 2-hydroxynicotinate, methyl 2-oxo-1,2-dihydro-3-pyridinecarboxylate, methyl 2-oxo-1,2-dihydropyridine-3-carboxylate, methyl 2-hydroxypyridine-3-carboxylate, 2-hydroxy-nicotinic acid methyl ester, 2-hydroxynicotinic acid methyl ester, 2-hydroxy-3-pyridinecarboxylic acid methyl ester, 3-pyridinecarboxylic acid, 2-hydroxy-, methyl ester, methyl 2-hydroxy-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 1,2-dihydro-2-oxo-, methyl ester
579028
methyl 2-oxo-1H-pyridine-3-carboxylate
COC(=O)C1=CC=CNC1=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 153°C to 155°C
Color Cream
Quantity 250 mg
Formula Weight 153.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 2-hydroxynicotinate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'