missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hexynoate, +98%, ACROS Organics™

Brand:  Acros Organics 382090050

Code : 29

Additional Details : CAS Number : 18937-79-6 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10259863

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Methyl 2-hexynoate
98+%
CH3(CH2)2C≡CCOOCH3
MFCD00041642
≥98%
C7H10O2
126.15

Specifications

Methyl 2-hexynoate
0.943
Clear colorless to yellow
18937-79-6
Authentic
1.4400 to 1.4420 (20°C, 589nm)
≥98%
C7H10O2
126.15
67°C
64.0° to 66.0°C (10.0 mmHg)
5g
0.9430g/mL
Glass bottle
Liquid
98+%
CH3(CH2)2C≡CCOOCH3
MFCD00041642
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl 2-hexynoate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS