missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-formylbenzoate, Thermo Scientific™, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | CH3OC(O)C6H4CHO |
Quantity | 10 g |
Formula Weight | 164.16 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 2-formylbenzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'