missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-fluorophenylacetate, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Specifications
Color | Colorless |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass Bottle |
Refractive Index | 1.4890 to 1.4910 |
Quantity | 100 g |
Formula Weight | 168.06 |
Percent Purity | 99% |
Physical Form | Liquid |
Chemical Name or Material | Methyl 2-fluorophenylacetate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'