missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-cyanoisonicotinate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Methyl 2-cyanoisonicotinate can be used in the synthesis of various pharmaceutical and biologically active compounds, such as tetra substituted imidazolines as potent and selective neuropeptide Y (NPY) Y5 receptor antagonists.
Solubility
Slightly Soluble in water (6.2 g/L) (25°C).
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Methyl 2-cyanoisonicotinate can be used in the synthesis of various pharmaceutical and biologically active compounds, such as tetra substituted imidazolines as potent and selective neuropeptide Y (NPY) Y5 receptor antagonists.
Solubility
Slightly Soluble in water (6.2 g/L) (25°C).
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Color | Brown |
Quantity | 1 g |
UN Number | UN3439 |
Solubility Information | Slightly Soluble in water (6.2g/L) (25°C). |
Formula Weight | 162.15 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | Methyl 2-cyanoisonicotinate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'