missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 2-cyanoisonicotinate, 97%

Brand:  Alfa Aesar™ H63090.06

Code : NEW

Additional Details : CAS Number : 94413-64-6 Weight : 0.00500kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15419588

  • £450.40 / 5g


Estimated Shipment date
from Supplier 23-03-2018
Add to basket

Description

Description

Applications
Methyl 2-cyanoisonicotinate can be used in the synthesis of various pharmaceutical and biologically active compounds, such as tetra substituted imidazolines as potent and selective neuropeptide Y (NPY) Y5 receptor antagonists.

Solubility
Slightly Soluble in water (6.2 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

Methyl 2-cyanoisonicotinate
162.15
UN3439
94413-64-6
MFCD07367894
Methyl 2-cyanopyridine-4-carboxylate; 2-Cyanopyridine-4-carboxylic acid methyl ester

Specifications

Methyl 2-cyanoisonicotinate
5g
Solid
162.15
UN3439
Pale brown
94413-64-6
97%
MFCD07367894
Methyl 2-cyanopyridine-4-carboxylate; 2-Cyanopyridine-4-carboxylic acid methyl ester
Videos
SDS