missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(chlorosulfonyl)benzoate, 90%, Tech., Thermo Scientific™

Catalog Number p-160924
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
26638-43-7
C8H7ClO4S
234.65
MFCD00009797
HUNUAFNLLYVTQD-UHFFFAOYSA-N
methyl 2-chlorosulfonyl benzoate, benzoic acid, 2-chlorosulfonyl-, methyl ester, unii-c6hzm8x6pk, c6hzm8x6pk, methyl 2-chlorosulphonyl benzoate, pubchem14808, acmc-1cirj, methyl 2-chiorosulfonylbenzoate, methyl 2 chlorosulfonyl benzoate, methyl2-chlorosulfonyl benzoate
117830
methyl 2-(chlorosulfonyl)benzoate
COC(=O)C1=CC=CC=C1S(Cl)(=O)=O
This item is not returnable. View return policy
26638-43-7
C8H7ClO4S
234.65
MFCD00009797
HUNUAFNLLYVTQD-UHFFFAOYSA-N
methyl 2-chlorosulfonyl benzoate, benzoic acid, 2-chlorosulfonyl-, methyl ester, unii-c6hzm8x6pk, c6hzm8x6pk, methyl 2-chlorosulphonyl benzoate, pubchem14808, acmc-1cirj, methyl 2-chiorosulfonylbenzoate, methyl 2 chlorosulfonyl benzoate, methyl2-chlorosulfonyl benzoate
117830
methyl 2-(chlorosulfonyl)benzoate
COC(=O)C1=CC=CC=C1S(Cl)(=O)=O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 58°C to 64°C
Color Beige to White or Pink
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula ClSO2C6H4COOCH3
Quantity 25 g
Beilstein 11, 373
Formula Weight 234.66
Percent Purity ≥88% (ex Cl)
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Methyl 2-(chlorosulfonyl)benzoate, tech., 90%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-(chlorosulfonyl)benzoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'