missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 2-bromothiazole-4-carboxylate, 96%

Brand:  Alfa Aesar™ H63541.03

Code : NEW

Additional Details : CAS Number : 170235-26-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15450279

  • £51.50 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Description

Description

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

Methyl 2-bromothiazole-4-carboxylate
222.06
2-Bromothiazole-4-carboxylic acid methyl ester
170235-26-4
MFCD06659908

Specifications

Methyl 2-bromothiazole-4-carboxylate
Solid
222.06
2-Bromothiazole-4-carboxylic acid methyl ester
170235-26-4
96%
MFCD06659908
1g
Videos
SDS