missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(bromomethyl)acrylate, 96%, Acros Organics™

Brand:  Acros Organics 367630050

Code : 29

Additional Details : CAS Number : 4224-69-5 Weight : 0.05500kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III


Quantity 5g

 View more versions of this product

Product Code. 10456442

  • £237.10 / 5g


Estimated Shipment
10-07-2018
Add to basket


Specifications

Chemical Identifiers

Methyl 2-(bromomethyl)acrylate
96%
179.01
96%
C5H7BrO2

Specifications

Methyl 2-(bromomethyl)acrylate
1.4890g/mL
Glass bottle
1.4870 to 1.4890
96%
1.489
Clear colorless to slightly brownish
78°C
4224-69-5
Authentic
Liquid
96%
C5H7BrO2
179.01
35.0° to 37.0°C (13.0 mmHg)
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl 2-(bromomethyl)acrylate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS