missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-bromobenzoate, 99%, Thermo Scientific Chemicals
19.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals
Solubility
Insoluble in water.
Notes
Store in a cool place. Keep away from oxidizing agents.
Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals
Solubility
Insoluble in water.
Notes
Store in a cool place. Keep away from oxidizing agents.
Specifications
Density | 1.532 |
Boiling Point | 252°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.559 |
Quantity | 25 g |
Beilstein | 1862501 |
Solubility Information | Insoluble in water. |
Formula Weight | 215.05 |
Percent Purity | 99% |
Chemical Name or Material | Methyl 2-bromobenzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'