missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 2-bromo-6-fluorophenylacetate, 96%

Brand:  Alfa Aesar™ H62465.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15478978

  • £476.30 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 2-bromo-6-fluorophenylacetate
247.06
2-Bromo-6-fluorophenylacetic acid methyl ester

Specifications

Methyl 2-bromo-6-fluorophenylacetate
Solid
247.06
5g
1427397-15-6
96%
2-Bromo-6-fluorophenylacetic acid methyl ester
Videos
SDS