missing translation for 'onlineSavingsMsg'
Learn More

(±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, 95%, Thermo Scientific Chemicals

Catalog Number 10282572
Click to view available options
5g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS 88568-95-0
CAS Min % 94.0
CAS Max % 100.0
Melting Point 77.0°C to 80.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C13H18NO7P
MDL Number MFCD00043304
Quantity 5 g
Synonym methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate, +/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, n-cbz-2-phosphonoglycine trimethyl ester, n-benzyloxy carbonyl, +/--cbz-alpha-phosphonoglycine trimethyl ester, methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate, +/--n-cbz-alpha-phosphonoglycine trimethyl ester, z-alpha-phosphonoglycine trimethyl ester, cbz-alpha-phosphono-dl-glycine trimethyl ester
InChI Key GSYSFVSGPABNNL-UHFFFAOYSA-N
SMILES COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
IUPAC Name methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate
Molecular Weight (g/mol) 331.26
PubChem CID 2734718
Formula Weight 331.26
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material (±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, 95%
Show More Show Less
compliance-icons
Product Identifier
  • (#1)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'