Methyl 2-aminonicotinate, 97%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: KM07908DE

5GR Methyl 2-aminonicotinate, 97%

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 14667-47-1 Weight: 0.05500kg Transport: UN number : 2811 Chem class : 1 Pack group : III


Packaging Amber Glass Bottle
Quantity 5g

Product Code. 10525574

Quantity Price
1 £ 43.0 / 5g
Estimated Shipment date
from Supplier 21-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material methyl 2-aminonicotinate
Assay 97%
Molecular Formula C7H8N2O2
Formula Weight 152.15

Specification

Formula Weight 152.15
Quantity 5g
Assay 97%
CAS 14667-47-1
Packaging Amber Glass Bottle
Molecular Formula C7H8N2O2
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material methyl 2-aminonicotinate
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Warning