missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-(trifluoromethyl)benzoate, 98%, Thermo Scientific Chemicals

Catalog Number 15488816
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
117324-58-0
C9H8F3NO2
219.163
MFCD08234902
QGFUDNJZHZNPCS-UHFFFAOYSA-N
14233808
methyl 2-amino-5-(trifluoromethyl)benzoate
COC(=O)C1=C(C=CC(=C1)C(F)(F)F)N
This item is not returnable. View return policy
117324-58-0
C9H8F3NO2
219.163
MFCD08234902
QGFUDNJZHZNPCS-UHFFFAOYSA-N
14233808
methyl 2-amino-5-(trifluoromethyl)benzoate
COC(=O)C1=C(C=CC(=C1)C(F)(F)F)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Synonym methyl 2-amino-5-trifluoromethyl benzoate, methyl2-amino-5-trifluoromethyl benzoate, 2-amino-5-trifluoromethyl benzoic acid methyl ester, benzoic acid,2-amino-5-trifluoromethyl-, methyl ester, benzoic acid, 2-amino-5-trifluoromethyl-, methyl ester, acmc-209z2m, benzoicacid,2-amino-5-trifluoromethyl-,methylester, methyl 5-trifluoromethylanthranilate, 2-amino-5-trifluoromethyl-benzoic acid methyl ester
Formula Weight 219.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-amino-5-(trifluoromethyl)benzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'