missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate, 95%, Thermo Scientific™

Catalog Number 15299706
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
184174-80-9
C9H11NO2S
197.25
MFCD00625612
XUKGZPUAFGAYHC-UHFFFAOYSA-N
methyl 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate, 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylic acid methyl ester, methyl 2-amino-4h,5h,6h-cyclopenta b thiophene-3-carboxylate, methyl 2-amino-5,6-dihydro-4h-cyclopenta b-thiophene-3-carboxylate, 4h-cyclopenta b thiophene-3-carboxylicacid, 2-amino-5,6-dihydro-, methyl ester, methyl 2-amino-4,5,6-trihydrocyclopenta 1,2-b thiophene-3-carboxylate, enamine_005617, 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-ca, methyl 2-azanyl-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate, 4h-cyclopenta b thiophene-3-carboxylic acid, 2-amino-5,6-dihydro-, methyl
2818378
methyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
COC(=O)C1=C(N)SC2=C1CCC2
This item is not returnable. View return policy
184174-80-9
C9H11NO2S
197.25
MFCD00625612
XUKGZPUAFGAYHC-UHFFFAOYSA-N
methyl 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate, 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylic acid methyl ester, methyl 2-amino-4h,5h,6h-cyclopenta b thiophene-3-carboxylate, methyl 2-amino-5,6-dihydro-4h-cyclopenta b-thiophene-3-carboxylate, 4h-cyclopenta b thiophene-3-carboxylicacid, 2-amino-5,6-dihydro-, methyl ester, methyl 2-amino-4,5,6-trihydrocyclopenta 1,2-b thiophene-3-carboxylate, enamine_005617, 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-ca, methyl 2-azanyl-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate, 4h-cyclopenta b thiophene-3-carboxylic acid, 2-amino-5,6-dihydro-, methyl
2818378
methyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
COC(=O)C1=C(N)SC2=C1CCC2
Melting Point 170°C to 174°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 197.26
Percent Purity 95%
Chemical Name or Material Methyl 2-amino-5, 6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
compliance-icons
Product Identifier
  • Methyl 2-amino-5,6-dihydro-4H-cyclopenta¢b!thiophene-3-carboxylate, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'