missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-4,5-difluorobenzoate, 98%, Thermo Scientific Chemicals

Catalog Number 15411579
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
207346-42-7
C8H7F2NO2
187.15
MFCD09264511
VIAQNTRKUPBQKR-UHFFFAOYSA-N
2-amino-4,5-difluorobenzoic acid methyl ester, benzoic acid, 2-amino-4,5-difluoro-, methyl ester, pubchem4676, acmc-209fdn, ksc496g7j, methyl 4,5-difluoroanthranilate, methyl2-amino-4,5-difluorobenzoate, methyl 2-amino-4,5-difluoro-benzoate, 2-amino-4,5-difluoro-benzoic acid methyl ester
18472001
methyl 2-amino-4,5-difluorobenzoate
COC(=O)C1=CC(F)=C(F)C=C1N
This item is not returnable. View return policy
207346-42-7
C8H7F2NO2
187.15
MFCD09264511
VIAQNTRKUPBQKR-UHFFFAOYSA-N
2-amino-4,5-difluorobenzoic acid methyl ester, benzoic acid, 2-amino-4,5-difluoro-, methyl ester, pubchem4676, acmc-209fdn, ksc496g7j, methyl 4,5-difluoroanthranilate, methyl2-amino-4,5-difluorobenzoate, methyl 2-amino-4,5-difluoro-benzoate, 2-amino-4,5-difluoro-benzoic acid methyl ester
18472001
methyl 2-amino-4,5-difluorobenzoate
COC(=O)C1=CC(F)=C(F)C=C1N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 100 g
Beilstein 2834936
Formula Weight 187.15
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-amino-4,5-difluorobenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'