missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-amino-3-carboxybenzoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 2-amino-3-methoxycarbonyl benzoic acid, methyl 2-amino-3-carboxybenzoate, 2-amino-isophthalic acid monomethyl ester, 3-methoxycarbonyl-2-aminobenzoic acid, 2-amino-isophthalicacidmonomethylester, 2-amino-3-methoxycarbonyl-benzoic acid, pubchem22030, 2-aminoisophthalicacidmonomethylester, 6-carboxy-2-methoxycarbonyl aniline, methyl2-amino-3-carboxybenzoate |
Formula Weight | 195.17 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 2-amino-3-carboxybenzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'