missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,6-dihydroxybenzoate, 97%, Thermo Scientific™

Catalog Number 11955101
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2150-45-0
C8H8O4
168.15
MFCD00059619
WCQZCKUNZVMBDC-UHFFFAOYSA-N
2,6-dihydroxybenzoic acid methyl ester, benzoic acid, 2,6-dihydroxy-, methyl ester, .gamma.-resorcylic acid, methyl ester, acmc-209fl9, 2-methoxycarbonyl resorcinol, methyl 2,6-dihydroxy benzoate, gamma-resorcylic acid methyl ester, methyl 2,6-dihydroxybenzoate, methyl-2,6-dihydroxybenzoate, 2-methoxycarbonyl benzene-1,3-diol
519869
methyl 2,6-dihydroxybenzoate
COC(=O)C1=C(O)C=CC=C1O
This item is not returnable. View return policy
2150-45-0
C8H8O4
168.15
MFCD00059619
WCQZCKUNZVMBDC-UHFFFAOYSA-N
2,6-dihydroxybenzoic acid methyl ester, benzoic acid, 2,6-dihydroxy-, methyl ester, .gamma.-resorcylic acid, methyl ester, acmc-209fl9, 2-methoxycarbonyl resorcinol, methyl 2,6-dihydroxy benzoate, gamma-resorcylic acid methyl ester, methyl 2,6-dihydroxybenzoate, methyl-2,6-dihydroxybenzoate, 2-methoxycarbonyl benzene-1,3-diol
519869
methyl 2,6-dihydroxybenzoate
COC(=O)C1=C(O)C=CC=C1O
Melting Point 67°C to 69°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material Methyl 2, 6-dihydroxybenzoate
compliance-icons
Product Identifier
  • Methyl 2,6-dihydroxybenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'