Learn More
Methyl 2,5-dihydroxycinnamate, Tocris Bioscience™
EGFR-kinase inhibitor
Brand:  Tocris (Bio-Techne) 057710
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13467773
  • £92.61 / 10mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Erbstatin analog. Inhibitor of EGF receptor-associated tyrosine kinases.
Specifications
Chemical Identifiers
63177-57-1
194.186
methyl 2,5-dihydroxycinnamate, erbstatin analog, methyl 3-2,5-dihydroxyphenyl acrylate, 2,5-dihydroxycinnamic acid methyl ester, 2,4-dihydroxymethylcinnamate, 2,5-dihydroxymethylcinnamate, chembl17329, methyl 3-2,5-dihydroxyphenyl-2-propenoate, methyl 2e-3-2,5-dihydroxyphenyl prop-2-enoate, 2-propenoic acid, 3-2,5-dihydroxyphenyl-, methyl ester
CHEBI:84089
COC(=O)C=CC1=C(C=CC(=C1)O)O
C10H10O4
BQCNSTFWSKOWMA-GORDUTHDSA-N
5353609
methyl (E)-3-(2,5-dihydroxyphenyl)prop-2-enoate
Specifications
10mg
63177-57-1
methyl 2,5-dihydroxycinnamate, erbstatin analog, methyl 3-2,5-dihydroxyphenyl acrylate, 2,5-dihydroxycinnamic acid methyl ester, 2,4-dihydroxymethylcinnamate, 2,5-dihydroxymethylcinnamate, chembl17329, methyl 3-2,5-dihydroxyphenyl-2-propenoate, methyl 2e-3-2,5-dihydroxyphenyl prop-2-enoate, 2-propenoic acid, 3-2,5-dihydroxyphenyl-, methyl ester
COC(=O)C=CC1=C(C=CC(=C1)O)O
194.186
CHEBI:84089
>98%
Methyl 2, 5-dihydroxycinnamate
C10H10O4
BQCNSTFWSKOWMA-GORDUTHDSA-N
methyl (E)-3-(2,5-dihydroxyphenyl)prop-2-enoate
5353609
194.19