missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate is a useful reagent for the trifluoromethylation of alkyl halides. It is also broadly applicable reagent for perfluoroalkylations for aryl iodides and bromides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate is a useful reagent for the trifluoromethylation of alkyl halides. It is also broadly applicable reagent for perfluoroalkylations for aryl iodides and bromides.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate is a useful reagent for the trifluoromethylation of alkyl halides. It is also broadly applicable reagent for perfluoroalkylations for aryl iodides and bromides.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications
Density | 1.509 |
Boiling Point | 117°C to 118°C |
Flash Point | 47°C (117°F) |
Linear Formula | FSO2CF2CO2CH3 |
Refractive Index | 1.351 |
Quantity | 5 g |
UN Number | UN2920 |
Beilstein | 1812896 |
Formula Weight | 192.11 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'