missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-methyl-5-oxopyrrolidine-3-carboxylate, 97%, Thermo Scientific™

Catalog Number 15418386
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
59857-86-2
C7H11NO3
157.17
MFCD00015898
JFZRRXGXEHUJCI-UHFFFAOYNA-N
108402
methyl 1-methyl-5-oxopyrrolidine-3-carboxylate
COC(=O)C1CN(C)C(=O)C1
This item is not returnable. View return policy
59857-86-2
C7H11NO3
157.17
MFCD00015898
JFZRRXGXEHUJCI-UHFFFAOYNA-N
108402
methyl 1-methyl-5-oxopyrrolidine-3-carboxylate
COC(=O)C1CN(C)C(=O)C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym methyl 1-methyl-2-oxopyrrolidine-4-carboxylate, 1-methyl-5-oxo-3-pyrrolidinecarboxylic acid methyl ester, 3-pyrrolidinecarboxylic acid, 1-methyl-5-oxo-, methyl ester, 1-methyl-4-methyloxycarbonyl-2-pyrrolidone, 1-methyl-5-oxo-pyrrolidine-3-carboxylic acid methyl ester, 5-22-06-00040 beilstein handbook reference, 4-carbomethoxy-1-methyl-pyrrolidone, methyl n-methyl-2-pyrrolidone-4-carboxylate, methyl1-methyl-2-oxopyrrolidine-4-carboxylate, methyl 1-methyl-5-oxo-3-pyrrolidinecarboxylate
Formula Weight 157.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 1-methyl-5-oxopyrrolidine-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'