missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-methyl-1H-imidazole-4-carboxylate, 97%, Thermo Scientific™

Catalog Number p-3076316
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
17289-19-9
C6H8N2O2
140.142
MFCD02179561
KZPZTVKOJSKVBV-UHFFFAOYSA-N
methyl 1-methyl-1h-imidazole-4-carboxylate, 1-methyl-1h-imidazole-4-carboxylic acid methyl ester, 1h-imidazole-4-carboxylic acid, 1-methyl-, methyl ester, pubchem9002, acmc-209e5g, methyl1-methyl-1h-imidazole-4-carboxylate, ksc534i0d, methyl 1-methylimidazol-4-carboxylate, methyl 1-methyl-1h-imidazole-carboxylate, 4-methoxycarbonyl-1-methyl-1h-imidazole
2773510
methyl 1-methylimidazole-4-carboxylate
CN1C=C(N=C1)C(=O)OC
This item is not returnable. View return policy
17289-19-9
C6H8N2O2
140.142
MFCD02179561
KZPZTVKOJSKVBV-UHFFFAOYSA-N
methyl 1-methyl-1h-imidazole-4-carboxylate, 1-methyl-1h-imidazole-4-carboxylic acid methyl ester, 1h-imidazole-4-carboxylic acid, 1-methyl-, methyl ester, pubchem9002, acmc-209e5g, methyl1-methyl-1h-imidazole-4-carboxylate, ksc534i0d, methyl 1-methylimidazol-4-carboxylate, methyl 1-methyl-1h-imidazole-carboxylate, 4-methoxycarbonyl-1-methyl-1h-imidazole
2773510
methyl 1-methylimidazole-4-carboxylate
CN1C=C(N=C1)C(=O)OC
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-imidazole-4-carboxylic acid methyl ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'