missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 1-benzyl-5-oxopyrrolidine-3-carboxylate, 98%

Brand:  Alfa Aesar™ H63564.03

Code : NEW

Additional Details : CAS Number : 51535-00-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15418376

  • £83.63 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 1-benzyl-5-oxopyrrolidine-3-carboxylate
233.26
1-Benzyl-5-oxopyrrolidine-3-carboxylic acid methyl ester
51535-00-3
MFCD00003196

Specifications

Methyl 1-benzyl-5-oxopyrrolidine-3-carboxylate
Solid
233.26
1-Benzyl-5-oxopyrrolidine-3-carboxylic acid methyl ester
1g
51535-00-3
98%
MFCD00003196
63° to 66°C
Videos
SDS