missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxypolyethylene glycol amine, M.W. 5,000, Thermo Scientific Chemicals
66.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Methoxypolyethylene glycol amine is used in the reduction of immunogenicity and increasing serum half life.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Granular Solid |
Solubility Information | Soluble in water,methanol,ethanol,chlorinated solvents and terahydrofuran. Insoluble in ether. |
Quantity | 1 g |
Chemical Name or Material | Methoxypolyethylene glycol amine, M.W. 5,000 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'