missing translation for 'onlineSavingsMsg'
Learn More

Methoxypolyethylene glycol amine, M.W. 1,000, Thermo Scientific Chemicals

Catalog Number p-7080146
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
80506-64-5
H2NCH2CH2(OCH2CH2)nOCH3
559.69
MFCD00131599
QWCGXANSAOXRFE-UHFFFAOYSA-N
2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine, mpeg12-nh2, mpeg12-amine, m-dpeg12-amine, m-peg12-amine, ambotzpeg1655, m-dpeg r 12-amine, aminopolyethylene glycol 5'000 monomethyl ether, 3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxaheptatriacontane-1-amine
2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine
COCCOCCN
This item is not returnable. View return policy
80506-64-5
H2NCH2CH2(OCH2CH2)nOCH3
559.69
MFCD00131599
QWCGXANSAOXRFE-UHFFFAOYSA-N
2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine, mpeg12-nh2, mpeg12-amine, m-dpeg12-amine, m-peg12-amine, ambotzpeg1655, m-dpeg r 12-amine, aminopolyethylene glycol 5'000 monomethyl ether, 3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxaheptatriacontane-1-amine
2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine
COCCOCCN

Methoxypolyethylene glycol amine is used in the reduction of immunogenicity and in increasing serum half life.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Granular Solid
Solubility Information Soluble in water,methanol,ethanol,chlorinated solvents and terahydrofuran. Insoluble in ether.
Quantity 250 mg
Chemical Name or Material Methoxypolyethylene glycol amine, M.W. 1,000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'