missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetyl chloride, 97%, stabilized, ACROS Organics™

Brand:  Acros Organics 125700500

Code : 29

Additional Details : CAS Number : 38870-89-2 Weight : 0.10000kg Transport : UN number : 2920 Chem class : 8 Pack group : II


Packaging Glass bottle
Quantity 50g

 View more versions of this product

Product Code. 10703013

  • £100.20 / 50g


Estimated Shipment
19-07-2018
Add to basket


Specifications

Chemical Identifiers

Methoxyacetyl chloride
38870-89-2
97%
CH3OCH2COCl
MFCD00000728
Stabilized
≥96.0% (RCOCl)
C3H5ClO2
108.52

Specifications

Methoxyacetyl chloride
38870-89-2
Authentic
1.4190 to 1.4240 (20°C, 589nm)
≥96.0% (RCOCl)
0.3%mgO
CH3OCH2COCl
MFCD00000728
Clear colorless to yellow
Stabilized
1.1870g/mL
Glass bottle
Liquid
97%
C3H5ClO2
108.52
12,310
50g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methoxyacetyl chloride, stabilized

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS