missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetyl chloride, 97%, stab. with ca 0.3% magnesium oxide, Thermo Scientific Chemicals

Catalog Number p-7058272
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
38870-89-2
C3H5ClO2
108.521
MFCD00000728
JJKWHOSQTYYFAE-UHFFFAOYSA-N
methoxyacetyl chloride, methoxyacetylchloride, acetyl chloride, methoxy, methoxy-acetyl chloride, methoxyacetic acid chloride, unii-t66s02b1s3, methoxy acetylchloride, 2-methoxyacetylchloride, methoxy acetyl chloride, ch3och2cocl
96623
2-methoxyacetyl chloride
COCC(=O)Cl
This item is not returnable. View return policy
38870-89-2
C3H5ClO2
108.521
MFCD00000728
JJKWHOSQTYYFAE-UHFFFAOYSA-N
methoxyacetyl chloride, methoxyacetylchloride, acetyl chloride, methoxy, methoxy-acetyl chloride, methoxyacetic acid chloride, unii-t66s02b1s3, methoxy acetylchloride, 2-methoxyacetylchloride, methoxy acetyl chloride, ch3och2cocl
96623
2-methoxyacetyl chloride
COCC(=O)Cl

Methoxyacetyl chloride is used as an intermediate for active pharmaceutical ingredients and dyes. Further, it acts as a precursor for the synthesis of agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxyacetyl chloride is used as an intermediate for active pharmaceutical ingredients and dyes. Further, it acts as a precursor for the synthesis of agrochemicals.

Solubility
Miscible with acetone, ether, carbon tetrachloride and chloroform.

Notes
Moisture sensitive. Store in a cool place. Incompatible with water, alcohols, oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point <-40°C
Density 1.185
Boiling Point 112°C to 113°C
Flash Point 28°C (82°F)
Refractive Index 1.424
Quantity 5 g
UN Number UN2920
Beilstein 1740244
Sensitivity Moisture sensitive
Solubility Information Miscible with acetone,ether,carbon tetrachloride and chloroform.
Formula Weight 108.52
Percent Purity 97%
Chemical Name or Material Methoxyacetyl chloride, stabilized with ≈0.3% magnesium oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'