Learn More
Methoxy(cyclooctadiene)iridium(I) dimer, Ir nominally 58%, Thermo Scientific Chemicals
Description
Methoxy(cyclooctadiene)iridium(I) dimer is used as a catalyst in the preparation of heteroaryl fused indole ring systems and borylation and Suzuki-miyaura coupling. Further, it is used in tetraborylation reactions, ortho-silylation of aryl ketone, benzaldehyde and benzyl alcohol derivatives through C-H activation. The most useful application is highly regio and enantio selection asymmetric hydroboration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Assay Percent Range | Ir nominally 58% |
Quantity | 5 g |
Sensitivity | Air, light, and moisture sensitive |
Formula Weight | 662.88 |
Concentration or Composition (by Analyte or Components) | Ir nominally 58% |
Physical Form | Powder |
Chemical Name or Material | Methoxy(cyclooctadiene)iridium(I) dimer |
RUO – Research Use Only