missing translation for 'onlineSavingsMsg'
Learn More

Methotrexate, Fisher BioReagents

Catalog Number p-27103
Click to view available options
Quantity:
10 mg
100 mg
Packaging:
Amber Glass
This item is not returnable. View return policy
59-05-2
C20H22N8O5
454.447
FBOZXECLQNJBKD-ZDUSSCGKSA-N
methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan
126941
CHEBI:44185
(2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O
This item is not returnable. View return policy
59-05-2
C20H22N8O5
454.447
FBOZXECLQNJBKD-ZDUSSCGKSA-N
methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan
126941
CHEBI:44185
(2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O
  • Inhibitor of DNA and RNA synthesis
  • Inhibits the enzyme responsible for synthesizing the folic acid derivative that is necessary for purine and pyrimidine synthesis
  • The lack of purines and pyrimidines  impacts the synthesis of DNA, RNA, and proteins
  • Referenced for use in antitumor studies and other studies of biological pathways
  • Intended for research use only
TRUSTED_SUSTAINABILITY
Decomposition Information >195°C
Melting Point 195°C
Color Yellow
Ignition Residue 0.1% max.
Assay Percent Range 98.0 to 102.0 %
Packaging Amber Glass
Quantity 10 mg
Merck Index 15, 6057
Specific Rotation +19.0 to +24.0° (+ or -)
Formula Weight 454.44
Percent Purity 98.0 to 102.0%
Physical Form Powder or Solid
Chemical Name or Material Methotrexate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methotrexate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H340-May cause genetic defects.
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'