missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals βmethasone, 97%
20.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Loss on Drying | 0.5% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Solubility Information | Solubility in water: slightly soluble. |
Formula Weight | 392.46 |
Percent Purity | 97% |
Chemical Name or Material | βmethasone |
Safety and Handling
Product Identifier
- Betamethasone
- Danger
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- H360D-May damage the unborn child.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'