missing translation for 'onlineSavingsMsg'
Learn More

Methanol, HPLC for Gradient Analysis, Fisher Chemical™

Catalog Number 10499560
41.63 GBP valid until 2024-12-31
Use promo code "21964" to get your promotional price.


38.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


38.04 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Plastic-coated Glass Bottle
67-56-1
CH4O
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
887
CHEBI:17790
methanol
CO
67-56-1
CH4O
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
887
CHEBI:17790
methanol
CO

Methanol, HPLC for Gradient Analysis

Specification
Molecular Formula: CH4O
Water Soluble: Yes
Molecular Weight: 32.04
Melting Point: -98°C
Health Hazards: Causes moderate eye irritation, may cause skin irritation, may cause irritation of the digestive tract
For Use With [Application]: HPLC for gradient analysis
Flash Point: 12°C
CAS No.: 67-56-1
Boiling Point: 64.7°C
Hazard Symbol: F, T
Synonyms: Carbinol, Methyl Alcohol, Methyl Hydroxide, Monohydroxymethane, Wood Alcohol, Wood Naptha, Wood Spirits, Columbian Spirits
TRUSTED_SUSTAINABILITY
Melting Point -98°C
Color Colorless
Boiling Point 64.7°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55cp at 20°C
Molecular Weight (g/mol) 32.042
Formula Weight 32.04g/mol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'