missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, HPLC for Gradient Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Plastic-coated Glass Bottle
Description
Methanol, HPLC for Gradient Analysis
Specification
Molecular Formula: CH4O
Water Soluble: Yes
Molecular Weight: 32.04
Melting Point: -98°C
Health Hazards: Causes moderate eye irritation, may cause skin irritation, may cause irritation of the digestive tract
For Use With [Application]: HPLC for gradient analysis
Flash Point: 12°C
CAS No.: 67-56-1
Boiling Point: 64.7°C
Hazard Symbol: F, T
Synonyms: Carbinol, Methyl Alcohol, Methyl Hydroxide, Monohydroxymethane, Wood Alcohol, Wood Naptha, Wood Spirits, Columbian Spirits
Molecular Formula: CH4O
Water Soluble: Yes
Molecular Weight: 32.04
Melting Point: -98°C
Health Hazards: Causes moderate eye irritation, may cause skin irritation, may cause irritation of the digestive tract
For Use With [Application]: HPLC for gradient analysis
Flash Point: 12°C
CAS No.: 67-56-1
Boiling Point: 64.7°C
Hazard Symbol: F, T
Synonyms: Carbinol, Methyl Alcohol, Methyl Hydroxide, Monohydroxymethane, Wood Alcohol, Wood Naptha, Wood Spirits, Columbian Spirits
Specifications
Melting Point | -98°C |
Color | Colorless |
Molecular Weight (g/mol) | 32.04 |
Formula Weight | 32.04g/mol |
Boiling Point | 64.7°C |
Quantity | 5 L |
Packaging | Aluminum Can |
Physical Form | Liquid |
Vapor Pressure | 128 hPa at 20°C |
Viscosity | 0.55cp at 20°C |
Safety and Handling
Product Identifier
- Methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P240-Ground/bond container and receiving equipment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'