missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Extra Dry, for Synthesis, Fisher Chemical™
222.00 GBP valid until 2024-12-31
Use promo code "21964" to get your promotional price.
Use promo code "21964" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Description
CH4O, CAS Number-67-56-1, methyl hydroxide, methylol, pyroxylic spirit, carbinol, wood spirit, wood naphtha, wood alcohol, colonial spirit, columbian spirit, methyl alcohol, 1L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042
Specifications
Melting Point | -98°C |
Color | Colorless |
Boiling Point | 64.7°C |
Quantity | 25 L |
Packaging | Metal Can |
Physical Form | Liquid |
Vapor Pressure | 128 hPa at 20°C |
Viscosity | 0.55cp at 20°C |
Formula Weight | 32.04g/mol |
Safety and Handling
Product Identifier
- Methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P240-Ground/bond container and receiving equipment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'